The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127713.1 | NP_001121185.1 | 4 - 5 | 840 | 97 |
NM_015915.4 | NP_056999.2 | 3 - 4 | 656 | 97 | |
NM_181598.3 | NP_853629.2 | 3 - 4 | 656 | 97 | |
GenBank mRNA | AF131801.1 | - | 2 - 3 | 180 | 97 |
AF444143.1 | - | 3 - 4 | 587 | 97 | |
AK290185.1 | - | 3 - 4 | 569 | 97 | |
AK312518.1 | - | 3 - 4 | 545 | 97 | |
AL833591.1 | - | 4 - 5 | 624 | 97 | |
AY032844.1 | - | 3 - 4 | 584 | 97 | |
BC010708.2 | - | 3 - 4 | 559 | 97 | |
CR457153.1 | - | 2 - 3 | 166 | 97 | |
KU178621.1 | - | 3 - 4 | 415 | 97 | |
KU178622.1 | - | 3 - 4 | 415 | 97 | |
KU178623.1 | - | 3 - 4 | 415 | 97 |