The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001004439.1 | NP_001004439.1 | 11 - 12 | 1362 | 91 |
XM_005254228.3 | XP_005254285.1 | 9 - 10 | 1584 | 91 | |
XM_011521363.2 | XP_011519665.1 | 9 - 10 | 1314 | 91 | |
GenBank mRNA | AF109681.1 | - | 11 - 12 | 1347 | 91 |
AF111799.1 | - | 3 - 4 | 327 | 91 | |
AF137378.2 | - | 11 - 12 | 1365 | 91 | |
BX496541.1 | - | 3 - 4 | 351 | 91 | |
CN358985.1 | - | 1 - 2 | 142 | 91 |