Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ITGA11
Entrez Gene ID
22801
Gene Name
integrin subunit alpha 11
Gene Aliases
HsT18964
Chromosome Location
Chr.15: 68297433 - 68432312 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004439.1 NP_001004439.1 20 - 21 2710 64
XM_005254228.3 XP_005254285.1 18 - 19 2932 64
XM_011521363.2 XP_011519665.1 18 - 19 2662 64
GenBank mRNA AF109681.1 - 20 - 21 2695 64
AF111799.1 - 12 - 13 1675 64
AF137378.2 - 20 - 21 2713 64
AK292445.1 - 20 - 21 2719 64

View Full Product Details