Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SYNDIG1L
Entrez Gene ID
646658
Gene Name
synapse differentiation inducing 1 like
Gene Aliases
CAPUCIN, DSPC1, IFITMD4, TMEM90A
Chromosome Location
Chr.14: 74405893 - 74477080 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105579.1 NP_001099049.1 3 - 4 801 70
XM_017021600.1 XP_016877089.1 3 - 4 3387 70
GenBank mRNA DB173128.1 - 2 - 3 293 70

View Full Product Details