The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025010.4 | NP_079286.2 | 6 - 7 | 1027 | 73 |
XM_005265003.3 | XP_005265060.2 | 6 - 7 | 927 | 73 | |
XM_017006035.1 | XP_016861524.1 | 5 - 6 | 790 | 73 | |
GenBank mRNA | AB018338.1 | - | 4 - 5 | 575 | 73 |
AB062478.1 | - | 6 - 7 | 1199 | 73 | |
AK023765.1 | - | 6 - 7 | 922 | 73 | |
AK291477.1 | - | 6 - 7 | 921 | 73 | |
AK295292.1 | - | 5 - 6 | 789 | 73 | |
BC016388.2 | - | 2 - 3 | 197 | 73 | |
BC032620.1 | - | 6 - 7 | 839 | 73 | |
BX537953.1 | - | 6 - 7 | 918 | 73 |