Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATL1
Entrez Gene ID
51062
Gene Name
atlastin GTPase 1
Gene Aliases
AD-FSP, FSP1, GBP3, HSN1D, SPG3, SPG3A, atlastin1
Chromosome Location
Chr.14: 50533082 - 50633068 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127713.1 NP_001121185.1 13 - 14 1972 107
NM_181598.3 NP_853629.2 12 - 13 1788 107
GenBank mRNA AL833591.1 - 13 - 14 1756 107
KU178622.1 - 12 - 13 1547 107

View Full Product Details