The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004080.2 | NP_004071.1 | 13 - 14 | 1357 | 74 |
NM_145695.2 | NP_663733.1 | 13 - 14 | 1357 | 74 | |
XM_005249628.2 | XP_005249685.1 | 14 - 15 | 1885 | 74 | |
XM_005249629.2 | XP_005249686.1 | 14 - 15 | 1882 | 74 | |
XM_005249630.2 | XP_005249687.1 | 13 - 14 | 1398 | 74 | |
XM_011515153.2 | XP_011513455.1 | 14 - 15 | 1723 | 74 | |
XM_011515154.2 | XP_011513456.1 | 14 - 15 | 1439 | 74 | |
XM_011515156.2 | XP_011513458.1 | 13 - 14 | 1395 | 74 | |
XM_017011783.1 | XP_016867272.1 | 14 - 15 | 1776 | 74 | |
XM_017011784.1 | XP_016867273.1 | 14 - 15 | 1436 | 74 | |
XM_017011785.1 | XP_016867274.1 | 13 - 14 | 1670 | 74 | |
XM_017011786.1 | XP_016867275.1 | 13 - 14 | 1650 | 74 | |
XM_017011787.1 | XP_016867276.1 | 13 - 14 | 1400 | 74 | |
XM_017011790.1 | XP_016867279.1 | 14 - 15 | 1722 | 74 | |
XM_017011791.1 | XP_016867280.1 | 14 - 15 | 1721 | 74 | |
XM_017011792.1 | XP_016867281.1 | 14 - 15 | 1721 | 74 | |
GenBank mRNA | AB018261.1 | - | 10 - 11 | 986 | 74 |
BC105005.1 | - | 13 - 14 | 1237 | 74 | |
BC143632.1 | - | 13 - 14 | 1234 | 74 |