The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008212.1 | NP_001008213.1 | 2 - 3 | 370 | 55 |
GenBank mRNA | AF420373.1 | - | 2 - 3 | 227 | 55 |
AK055403.1 | - | 2 - 3 | 659 | 55 |