The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033025.2 | NP_001028197.1 | 3 - 4 | 1050 | 83 |
NM_001261441.1 | NP_001248370.1 | 4 - 5 | 1074 | 83 | |
NM_001439.3 | NP_001430.1 | 3 - 4 | 1086 | 83 | |
NR_048570.1 | 4 - 5 | 1181 | 83 | ||
XM_005270621.1 | XP_005270678.1 | 4 - 5 | 916 | 83 | |
XM_011540994.2 | XP_011539296.1 | 4 - 5 | 1342 | 83 | |
XM_011540995.2 | XP_011539297.1 | 5 - 6 | 1359 | 83 | |
GenBank mRNA | AB009284.1 | - | 3 - 4 | 719 | 83 |
AF000416.1 | - | 3 - 4 | 719 | 83 | |
AK298124.1 | - | 4 - 5 | 834 | 83 | |
AK312895.1 | - | 3 - 4 | 655 | 83 | |
AU130571.1 | - | 4 - 5 | 559 | 83 | |
BC014910.1 | - | 3 - 4 | 606 | 83 | |
BC036015.1 | - | 3 - 4 | 696 | 83 | |
BP202626.1 | - | 1 - 2 | 387 | 83 |