The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001316345.1 | NP_001303274.1 | 9 - 10 | 661 | 72 |
NM_001316346.1 | NP_001303275.1 | 7 - 8 | 525 | 72 | |
NM_001316347.1 | NP_001303276.1 | 7 - 8 | 525 | 72 | |
NM_001316348.1 | NP_001303277.1 | 7 - 8 | 525 | 72 | |
NM_021111.2 | NP_066934.1 | 7 - 8 | 525 | 72 | |
XM_017015207.1 | XP_016870696.1 | 8 - 9 | 561 | 72 | |
XM_017015208.1 | XP_016870697.1 | 8 - 9 | 635 | 72 | |
XM_017015209.1 | XP_016870698.1 | 8 - 9 | 624 | 72 | |
XM_017015210.1 | XP_016870699.1 | 6 - 7 | 376 | 72 | |
XM_017015211.1 | XP_016870700.1 | 7 - 8 | 527 | 72 | |
GenBank mRNA | AK292653.1 | - | 7 - 8 | 525 | 72 |
AK315073.1 | - | 7 - 8 | 511 | 72 | |
BC060806.1 | - | 7 - 8 | 499 | 72 | |
BC137093.1 | - | 7 - 8 | 486 | 72 | |
BX648668.1 | - | 5 - 6 | 403 | 72 | |
D50406.1 | - | 7 - 8 | 548 | 72 | |
JQ756320.1 | - | 7 - 8 | 439 | 72 |