Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RECK
Entrez Gene ID
8434
Gene Name
reversion inducing cysteine rich protein with kazal motifs
Gene Aliases
ST15
Chromosome Location
Chr.9: 36036905 - 36124455 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001316345.1 NP_001303274.1 11 - 12 1129 109
NM_021111.2 NP_066934.1 9 - 10 993 109
XM_017015207.1 XP_016870696.1 10 - 11 1029 109
XM_017015208.1 XP_016870697.1 10 - 11 1103 109
XM_017015209.1 XP_016870698.1 10 - 11 1092 109
XM_017015210.1 XP_016870699.1 8 - 9 844 109
XM_017015211.1 XP_016870700.1 9 - 10 995 109
GenBank mRNA AK292653.1 - 9 - 10 993 109
AK315073.1 - 9 - 10 979 109
BC137093.1 - 9 - 10 954 109
BX648668.1 - 7 - 8 871 109
D50406.1 - 9 - 10 1016 109

View Full Product Details