Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAH6
Entrez Gene ID
1768
Gene Name
dynein axonemal heavy chain 6
Gene Aliases
DNHL1, Dnahc6, HL-2, HL2
Chromosome Location
Chr.2: 84459588 - 84819589 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001370.1 NP_001361.1 34 - 35 5773 77
XM_006711956.2 XP_006712019.1 34 - 35 5878 77
XM_011532649.2 XP_011530951.1 34 - 35 5727 77
XM_011532650.2 XP_011530952.1 34 - 35 5728 77
XM_011532652.1 XP_011530954.1 33 - 34 5647 77
XM_011532653.1 XP_011530955.1 34 - 35 5773 77
XM_011532654.1 XP_011530956.1 33 - 34 5506 77
XM_011532655.2 XP_011530957.1 30 - 31 4795 77
XM_011532657.1 XP_011530959.1 28 - 29 4534 77
XM_011532660.1 XP_011530962.1 34 - 35 5773 77
XM_017003521.1 XP_016859010.1 33 - 34 5647 77
XM_017003522.1 XP_016859011.1 29 - 30 5599 77
XM_017003523.1 XP_016859012.1 29 - 30 4766 77
XM_017003524.1 XP_016859013.1 34 - 35 5773 77

View Full Product Details