Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAH6
Entrez Gene ID
1768
Gene Name
dynein axonemal heavy chain 6
Gene Aliases
DNHL1, Dnahc6, HL-2, HL2
Chromosome Location
Chr.2: 84459588 - 84819589 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001370.1 NP_001361.1 35 - 36 5897 158
XM_006711956.2 XP_006712019.1 35 - 36 6002 158
XM_011532649.2 XP_011530951.1 35 - 36 5851 158
XM_011532650.2 XP_011530952.1 35 - 36 5852 158
XM_011532652.1 XP_011530954.1 34 - 35 5771 158
XM_011532653.1 XP_011530955.1 35 - 36 5897 158
XM_011532654.1 XP_011530956.1 34 - 35 5630 158
XM_011532655.2 XP_011530957.1 31 - 32 4919 158
XM_011532657.1 XP_011530959.1 29 - 30 4658 158
XM_011532660.1 XP_011530962.1 35 - 36 5897 158
XM_017003521.1 XP_016859010.1 34 - 35 5771 158
XM_017003522.1 XP_016859011.1 30 - 31 5723 158
XM_017003523.1 XP_016859012.1 30 - 31 4890 158
XM_017003524.1 XP_016859013.1 35 - 36 5897 158

View Full Product Details