The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001370.1 | NP_001361.1 | 60 - 61 | 10110 | 111 |
XM_006711956.2 | XP_006712019.1 | 60 - 61 | 10215 | 111 | |
XM_011532649.2 | XP_011530951.1 | 60 - 61 | 10064 | 111 | |
XM_011532650.2 | XP_011530952.1 | 60 - 61 | 10065 | 111 | |
XM_011532652.1 | XP_011530954.1 | 59 - 60 | 9984 | 111 | |
XM_011532653.1 | XP_011530955.1 | 59 - 60 | 9963 | 111 | |
XM_011532654.1 | XP_011530956.1 | 59 - 60 | 9843 | 111 | |
XM_011532655.2 | XP_011530957.1 | 56 - 57 | 9132 | 111 | |
XM_011532657.1 | XP_011530959.1 | 54 - 55 | 8871 | 111 | |
XM_017003521.1 | XP_016859010.1 | 58 - 59 | 9837 | 111 | |
XM_017003522.1 | XP_016859011.1 | 55 - 56 | 9936 | 111 | |
XM_017003523.1 | XP_016859012.1 | 55 - 56 | 9103 | 111 | |
GenBank mRNA | AB051484.2 | - | 24 - 25 | 4045 | 111 |
AB290165.1 | - | 26 - 27 | 4249 | 111 | |
AL512706.1 | - | 2 - 3 | 405 | 111 | |
BC117259.1 | - | 13 - 14 | 2242 | 111 |