The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001370.1 | NP_001361.1 | 46 - 47 | 7661 | 118 |
XM_006711956.2 | XP_006712019.1 | 46 - 47 | 7766 | 118 | |
XM_011532649.2 | XP_011530951.1 | 46 - 47 | 7615 | 118 | |
XM_011532650.2 | XP_011530952.1 | 46 - 47 | 7616 | 118 | |
XM_011532652.1 | XP_011530954.1 | 45 - 46 | 7535 | 118 | |
XM_011532653.1 | XP_011530955.1 | 45 - 46 | 7514 | 118 | |
XM_011532654.1 | XP_011530956.1 | 45 - 46 | 7394 | 118 | |
XM_011532655.2 | XP_011530957.1 | 42 - 43 | 6683 | 118 | |
XM_011532657.1 | XP_011530959.1 | 40 - 41 | 6422 | 118 | |
XM_017003521.1 | XP_016859010.1 | 44 - 45 | 7388 | 118 | |
XM_017003522.1 | XP_016859011.1 | 41 - 42 | 7487 | 118 | |
XM_017003523.1 | XP_016859012.1 | 41 - 42 | 6654 | 118 | |
GenBank mRNA | AB051484.2 | - | 10 - 11 | 1596 | 118 |
AB290165.1 | - | 12 - 13 | 1800 | 118 | |
BC015442.1 | - | 10 - 11 | 1605 | 118 |