The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017647.3 | NP_060117.3 | 7 - 8 | 1244 | 115 |
GenBank mRNA | AF327355.1 | - | 6 - 7 | 719 | 115 |
AK095861.1 | - | 7 - 8 | 1244 | 115 | |
AK307398.1 | - | 6 - 7 | 733 | 115 | |
AK315010.1 | - | 7 - 8 | 669 | 115 | |
AL834482.1 | - | 1 - 2 | 132 | 115 | |
BC000131.1 | - | 2 - 3 | 259 | 115 | |
BC036710.2 | - | 6 - 7 | 628 | 115 |