Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZNF44
Entrez Gene ID
51710
Gene Name
zinc finger protein 44
Gene Aliases
GIOT-2, KOX7, ZNF, ZNF504, ZNF55, ZNF58
Chromosome Location
Chr.19: 12224686 - 12294904 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AB021642.1 - 5 - 6 2027 103
BC136502.1 - 6 - 7 2131 103

View Full Product Details