Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ZNF44
- Entrez Gene ID
-
51710
- Gene Name
- zinc finger protein 44
- Gene Aliases
-
GIOT-2, KOX7, ZNF, ZNF504, ZNF55, ZNF58
- Chromosome Location
-
Chr.19: 12224686 - 12294904 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC032246.1 |
- |
10 - 11 |
1301 |
130 |
- Gene Symbol
- LOC101928574
- Entrez Gene ID
-
101928574
- Gene Name
- uncharacterized LOC101928574
- Gene Aliases
-
-
- Chromosome Location
-
Chr.19: 12207435 - 12242746 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AL137602.1 |
- |
3 - 4 |
337 |
130 |
View Full Product Details