Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZNF44
Entrez Gene ID
51710
Gene Name
zinc finger protein 44
Gene Aliases
GIOT-2, KOX7, ZNF, ZNF504, ZNF55, ZNF58
Chromosome Location
Chr.19: 12224686 - 12294904 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164276.1 NP_001157748.1 2 - 3 349 72
XM_017026871.1 XP_016882360.1 2 - 3 343 72
GenBank mRNA AK297528.1 - 2 - 3 263 72
BC032246.1 - 2 - 3 272 72
BC136502.1 - 2 - 3 244 72

View Full Product Details