The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032221.4 | NP_115597.3 | 21 - 22 | 3503 | 71 |
XM_005260573.2 | XP_005260630.1 | 20 - 21 | 3325 | 71 | |
XM_011529080.2 | XP_011527382.1 | 20 - 21 | 3564 | 71 | |
XM_011529082.2 | XP_011527384.1 | 10 - 11 | 1736 | 71 | |
XM_017028099.1 | XP_016883588.1 | 20 - 21 | 4999 | 71 | |
XM_017028100.1 | XP_016883589.1 | 20 - 21 | 3524 | 71 | |
XM_017028101.1 | XP_016883590.1 | 19 - 20 | 4855 | 71 | |
XM_017028102.1 | XP_016883591.1 | 20 - 21 | 4998 | 71 | |
XM_017028103.1 | XP_016883592.1 | 18 - 19 | 2958 | 71 | |
XM_017028104.1 | XP_016883593.1 | 20 - 21 | 4999 | 71 | |
GenBank mRNA | AB037756.2 | - | 7 - 8 | 1304 | 71 |
AK026022.1 | - | 6 - 7 | 1052 | 71 | |
AY034072.1 | - | 21 - 22 | 3503 | 71 |