Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHD6
Entrez Gene ID
84181
Gene Name
chromodomain helicase DNA binding protein 6
Gene Aliases
CHD-6, CHD5, RIGB
Chromosome Location
Chr.20: 41402101 - 41618494 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032221.4 NP_115597.3 28 - 29 4311 87
XM_005260573.2 XP_005260630.1 27 - 28 4133 87
XM_011529080.2 XP_011527382.1 27 - 28 4372 87
XM_011529082.2 XP_011527384.1 17 - 18 2544 87
XM_017028099.1 XP_016883588.1 27 - 28 5807 87
XM_017028100.1 XP_016883589.1 27 - 28 4332 87
XM_017028101.1 XP_016883590.1 26 - 27 5663 87
XM_017028102.1 XP_016883591.1 27 - 28 5806 87
XM_017028103.1 XP_016883592.1 25 - 26 3766 87
XM_017028104.1 XP_016883593.1 27 - 28 5807 87
GenBank mRNA AB037756.2 - 14 - 15 2112 87
AK026022.1 - 13 - 14 1860 87
AK091551.1 - 15 - 16 2216 87
AY034072.1 - 28 - 29 4311 87

View Full Product Details