The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024949.5 | NP_079225.5 | 19 - 20 | 3225 | 62 |
XM_011532269.2 | XP_011530571.1 | 19 - 20 | 3930 | 62 | |
GenBank mRNA | AJ566366.1 | - | 6 - 7 | 907 | 62 |
AK025682.1 | - | 1 - 2 | 155 | 62 | |
AK074260.1 | - | 6 - 7 | 937 | 62 | |
AK126057.1 | - | 20 - 21 | 3119 | 62 | |
AK127061.1 | - | 12 - 13 | 2137 | 62 | |
AK225323.1 | - | 1 - 2 | 155 | 62 | |
BC017957.1 | - | 2 - 3 | 460 | 62 | |
BC053873.1 | - | 10 - 11 | 1654 | 62 | |
BX647378.1 | - | 17 - 18 | 2816 | 62 | |
BX647704.1 | - | 7 - 8 | 1024 | 62 |