Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
XRCC3
Entrez Gene ID
7517
Gene Name
X-ray repair cross complementing 3
Gene Aliases
CMM6
Chromosome Location
Chr.14: 103697611 - 103715486 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005268046.2 XP_005268103.1 1 - 2 67 77

View Full Product Details