The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080742.2 | NP_542780.1 | 1 - 2 | 1252 | 73 |
GenBank mRNA | AB075843.1 | - | 1 - 2 | 1205 | 73 |
AK055248.1 | - | 1 - 2 | 128 | 73 | |
AK290018.1 | - | 1 - 2 | 152 | 73 | |
AY070019.1 | - | 1 - 2 | 596 | 73 | |
BC101163.2 | - | 2 - 3 | 236 | 73 | |
BC101165.2 | - | 2 - 3 | 245 | 73 |