Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FMO2
Entrez Gene ID
2327
Gene Name
flavin containing monooxygenase 2
Gene Aliases
FMO1B1
Chromosome Location
Chr.1: 171185208 - 171212683 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001460.4 NP_001451.2 5 - 6 781 91
GenBank mRNA AK312877.1 - 5 - 6 687 91
AL833218.1 - 5 - 6 670 91
BC005894.1 - 5 - 6 687 91
BT006979.1 - 4 - 5 623 91
Y09267.1 - 5 - 6 683 91

View Full Product Details