The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014663.2 | NP_055478.2 | 12 - 13 | 2020 | 76 |
XM_005271354.3 | XP_005271411.1 | 12 - 13 | 2057 | 76 | |
XM_005271355.3 | XP_005271412.1 | 12 - 13 | 1975 | 76 | |
XM_005271356.3 | XP_005271413.1 | 13 - 14 | 2096 | 76 | |
XM_011542467.2 | XP_011540769.2 | 3 - 4 | 1128 | 76 | |
XM_017002912.1 | XP_016858401.1 | 13 - 14 | 2165 | 76 | |
GenBank mRNA | AB014577.1 | - | 12 - 13 | 1979 | 76 |
BC002558.2 | - | 12 - 13 | 1988 | 76 |