The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014663.2 | NP_055478.2 | 2 - 3 | 299 | 77 |
XM_005271354.3 | XP_005271411.1 | 2 - 3 | 336 | 77 | |
XM_005271355.3 | XP_005271412.1 | 2 - 3 | 254 | 77 | |
GenBank mRNA | AB014577.1 | - | 2 - 3 | 258 | 77 |
AK300039.1 | - | 2 - 3 | 291 | 77 | |
BC002558.2 | - | 2 - 3 | 267 | 77 | |
BP198064.1 | - | 2 - 3 | 299 | 77 |