The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020421.3 | NP_065154.2 | 4 - 5 | 521 | 61 |
XM_005267885.3 | XP_005267942.1 | 4 - 5 | 544 | 61 | |
XM_005267886.3 | XP_005267943.1 | 4 - 5 | 540 | 61 | |
XM_011536987.2 | XP_011535289.1 | 4 - 5 | 540 | 61 | |
XM_011536988.2 | XP_011535290.1 | 4 - 5 | 540 | 61 | |
XM_017021474.1 | XP_016876963.1 | 4 - 5 | 540 | 61 | |
XM_017021475.1 | XP_016876964.1 | 4 - 5 | 540 | 61 | |
XM_017021476.1 | XP_016876965.1 | 5 - 6 | 502 | 61 | |
XM_017021477.1 | XP_016876966.1 | 2 - 3 | 219 | 61 | |
GenBank mRNA | BC017459.1 | - | 4 - 5 | 478 | 61 |
BC058906.1 | - | 4 - 5 | 473 | 61 | |
BX248292.1 | - | 4 - 5 | 504 | 61 |