The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005483.2 | NP_005474.2 | 12 - 13 | 2301 | 89 |
XM_011527605.2 | XP_011525907.1 | 12 - 13 | 2368 | 89 | |
XM_011527606.2 | XP_011525908.1 | 12 - 13 | 2368 | 89 | |
XM_011527607.2 | XP_011525909.1 | 11 - 12 | 2311 | 89 | |
GenBank mRNA | BC067093.1 | - | 12 - 13 | 2302 | 89 |
BG775588.1 | - | 1 - 2 | 183 | 89 | |
U20979.1 | - | 12 - 13 | 2187 | 89 |