The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142301.1 | NP_001135773.1 | 10 - 11 | 975 | 154 |
NM_153704.5 | NP_714915.3 | 9 - 10 | 1044 | 154 | |
NR_024522.1 | 9 - 10 | 1044 | 154 | ||
XM_006716686.3 | XP_006716749.1 | 8 - 9 | 882 | 154 | |
GenBank mRNA | AK026742.1 | - | 4 - 5 | 349 | 155 |
AK094935.1 | - | 9 - 10 | 968 | 154 | |
AK308071.1 | - | 10 - 11 | 934 | 154 | |
BC032835.2 | - | 9 - 10 | 962 | 154 | |
BC054338.1 | - | 9 - 10 | 971 | 154 | |
BX648768.1 | - | 9 - 10 | 988 | 154 | |
KU178827.1 | - | 9 - 10 | 943 | 154 | |
KU178828.1 | - | 8 - 9 | 789 | 154 |