Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYTH3
Entrez Gene ID
9265
Gene Name
cytohesin 3
Gene Aliases
ARNO3, GRP1, PSCD3
Chromosome Location
Chr.7: 6161779 - 6272611 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004227.3 NP_004218.1 5 - 6 471 117
GenBank mRNA AJ005197.1 - 5 - 6 442 117
AJ223957.1 - 5 - 6 392 117
AK313728.1 - 5 - 6 487 117
BC028717.1 - 5 - 6 507 117
CB988199.1 - 5 - 6 503 117

View Full Product Details