Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PIANP
Entrez Gene ID
196500
Gene Name
PILR alpha associated neural protein
Gene Aliases
C12orf53, LEDA1, PANP, leda-1
Chromosome Location
Chr.12: 6689726 - 6700843 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244014.1 NP_001230943.1 2 - 3 292 93
NM_001244015.1 NP_001230944.1 2 - 3 358 93
NM_153685.3 NP_710152.1 2 - 3 292 93
XM_011520926.2 XP_011519228.1 2 - 3 249 93
GenBank mRNA AK056295.1 - 2 - 3 249 93
AK091596.1 - 2 - 3 92 93
AK095854.1 - 2 - 3 358 93
AK289648.1 - 2 - 3 147 93
AY358534.1 - 1 - 2 76 93
BC035736.1 - 2 - 3 95 93

View Full Product Details