The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318798.1 | NP_001305727.1 | 13 - 14 | 1486 | 127 |
NM_001318800.1 | NP_001305729.1 | 13 - 14 | 1770 | 127 | |
NM_004186.4 | NP_004177.3 | 14 - 15 | 1747 | 127 | |
XM_005265381.4 | XP_005265438.1 | 14 - 15 | 1877 | 127 | |
XM_005265382.4 | XP_005265439.1 | 13 - 14 | 1587 | 127 | |
XM_006713289.3 | XP_006713352.1 | 8 - 9 | 1359 | 127 | |
XM_011533998.2 | XP_011532300.1 | 14 - 15 | 1679 | 127 | |
GenBank mRNA | AB209259.1 | - | 13 - 14 | 1440 | 127 |
BC042914.1 | - | 14 - 15 | 1743 | 127 | |
U33920.1 | - | 13 - 14 | 1623 | 127 | |
U38276.1 | - | 13 - 14 | 1764 | 127 |