The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282409.1 | NP_001269338.1 | 4 - 5 | 1141 | 108 |
NM_001282410.1 | NP_001269339.1 | 5 - 6 | 1270 | 108 | |
NM_001282411.1 | NP_001269340.1 | 4 - 5 | 1141 | 108 | |
NM_001282412.1 | NP_001269341.1 | 5 - 6 | 516 | 108 | |
NM_001282413.1 | NP_001269342.1 | 4 - 5 | 387 | 108 | |
NM_001282414.1 | NP_001269343.1 | 4 - 5 | 387 | 108 | |
NM_002712.2 | NP_002703.1 | 5 - 6 | 1270 | 108 | |
XM_011511388.1 | XP_011509690.1 | 4 - 5 | 485 | 108 | |
GenBank mRNA | AK294043.1 | - | 4 - 5 | 342 | 108 |
AK308015.1 | - | 5 - 6 | 481 | 108 | |
BC000910.2 | - | 5 - 6 | 443 | 108 | |
BC012397.1 | - | 5 - 6 | 436 | 108 | |
BC013001.2 | - | 5 - 6 | 448 | 108 | |
BG684153.1 | - | 4 - 5 | 313 | 108 | |
BI256145.1 | - | 4 - 5 | 301 | 108 | |
BQ212238.1 | - | 4 - 5 | 368 | 108 | |
BT007296.1 | - | 5 - 6 | 432 | 108 | |
BT020134.1 | - | 5 - 6 | 432 | 108 | |
Z50749.1 | - | 5 - 6 | 447 | 108 |