The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199282.2 | NP_001186211.2 | 18 - 19 | 2545 | 111 |
NM_006726.4 | NP_006717.2 | 18 - 19 | 2504 | 111 | |
XM_005263372.3 | XP_005263429.1 | 18 - 19 | 2690 | 111 | |
XM_005263373.2 | XP_005263430.1 | 18 - 19 | 2732 | 111 | |
XM_005263374.3 | XP_005263431.1 | 18 - 19 | 2689 | 111 | |
XM_005263375.3 | XP_005263432.1 | 18 - 19 | 2689 | 111 | |
XM_011532434.2 | XP_011530736.1 | 18 - 19 | 2689 | 111 | |
XM_017008872.1 | XP_016864361.1 | 18 - 19 | 2732 | 111 | |
GenBank mRNA | AF216648.2 | - | 18 - 19 | 111 | |
AF467287.1 | - | 18 - 19 | 2545 | 111 |