Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRBA
Entrez Gene ID
987
Gene Name
LPS responsive beige-like anchor protein
Gene Aliases
BGL, CDC4L, CVID8, LAB300, LBA
Chromosome Location
Chr.4: 150264515 - 151015725 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199282.2 NP_001186211.2 43 - 44 6955 106
NM_006726.4 NP_006717.2 44 - 45 6947 106
XM_005263372.3 XP_005263429.1 44 - 45 7133 106
XM_005263373.2 XP_005263430.1 44 - 45 7175 106
XM_005263374.3 XP_005263431.1 43 - 44 7099 106
XM_005263375.3 XP_005263432.1 43 - 44 7099 106
XM_011532434.2 XP_011530736.1 44 - 45 7132 106
XM_017008872.1 XP_016864361.1 43 - 44 7142 106
XM_017008873.1 XP_016864362.1 4 - 5 857 106
XM_017008874.1 XP_016864363.1 4 - 5 563 106
GenBank mRNA AB208838.1 - 19 - 20 2628 106
AF216648.2 - 44 - 45 6947 106
AF467287.1 - 43 - 44 6955 106
AK122920.1 - 7 - 8 931 106
CR749633.1 - 4 - 5 745 106
M83822.1 - 26 - 27 4383 106

View Full Product Details