Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRBA
Entrez Gene ID
987
Gene Name
LPS responsive beige-like anchor protein
Gene Aliases
BGL, CDC4L, CVID8, LAB300, LBA
Chromosome Location
Chr.4: 150264515 - 151015725 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199282.2 NP_001186211.2 51 - 52 7984 62
NM_006726.4 NP_006717.2 52 - 53 7976 62
XM_005263372.3 XP_005263429.1 52 - 53 8177 62
XM_005263373.2 XP_005263430.1 52 - 53 8219 62
XM_005263374.3 XP_005263431.1 51 - 52 8143 62
XM_005263375.3 XP_005263432.1 51 - 52 8128 62
XM_011532434.2 XP_011530736.1 52 - 53 8161 62
XM_017008872.1 XP_016864361.1 51 - 52 8186 62
XM_017008873.1 XP_016864362.1 12 - 13 1901 62
XM_017008874.1 XP_016864363.1 12 - 13 1607 62
GenBank mRNA AB208838.1 - 27 - 28 3657 62
AF216648.2 - 52 - 53 7976 62
AF217149.1 - 5 - 6 568 62
AF467287.1 - 51 - 52 7984 62
AK026483.1 - 5 - 6 529 62
AK122920.1 - 15 - 16 1960 62
CR749633.1 - 12 - 13 1789 62
M83822.1 - 34 - 35 5412 62

View Full Product Details