Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- UBA1
- Entrez Gene ID
-
7317
- Gene Name
- ubiquitin like modifier activating enzyme 1
- Gene Aliases
-
A1S9, A1S9T, A1ST, AMCX1, CFAP124, GXP1, POC20, SMAX2, UBA1A, UBE1, UBE1X
- Chromosome Location
-
Chr.X: 47190800 - 47215128 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_153280.2 |
NP_695012.1 |
1 - 2 |
151 |
85 |
GenBank mRNA |
BC013041.2 |
- |
1 - 2 |
90 |
85 |
View Full Product Details