The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024086.3 | NP_076991.3 | 6 - 7 | 879 | 111 |
XM_005256801.2 | XP_005256858.1 | 5 - 6 | 716 | 111 | |
XM_017025045.1 | XP_016880534.1 | 7 - 8 | 922 | 111 | |
XM_017025046.1 | XP_016880535.1 | 4 - 5 | 630 | 111 | |
GenBank mRNA | AK027410.1 | - | 6 - 7 | 859 | 111 |
AK294547.1 | - | 5 - 6 | 692 | 111 | |
AK296464.1 | - | 4 - 5 | 604 | 111 | |
AL832612.2 | - | 6 - 7 | 838 | 111 | |
BC050603.1 | - | 6 - 7 | 819 | 111 | |
BX647255.1 | - | 5 - 6 | 653 | 111 |