The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000189.4 | NP_000180.2 | 1 - 2 | 1985 | 63 |
XM_005264280.1 | XP_005264337.1 | 1 - 2 | 465 | 63 | |
XM_011532807.1 | XP_011531109.1 | 1 - 2 | 465 | 63 | |
XM_017003945.1 | XP_016859434.1 | 1 - 2 | 465 | 63 | |
GenBank mRNA | AF148513.1 | - | 1 - 2 | 508 | 63 |
AK290357.1 | - | 1 - 2 | 488 | 63 | |
BC021116.1 | - | 1 - 2 | 503 | 63 | |
BC064369.1 | - | 1 - 2 | 501 | 63 |