The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005189.2 | NP_005180.1 | 3 - 4 | 246 | 62 |
NM_032647.3 | NP_116036.1 | 3 - 4 | 246 | 62 | |
XM_011525382.1 | XP_011523684.1 | 3 - 4 | 286 | 62 | |
XM_011525383.2 | XP_011523685.1 | 3 - 4 | 1248 | 62 | |
GenBank mRNA | BC004252.1 | - | 3 - 4 | 230 | 62 |
BC119759.1 | - | 3 - 4 | 217 | 62 | |
BC119760.1 | - | 3 - 4 | 217 | 62 | |
CN367085.1 | - | 3 - 4 | 247 | 62 | |
CN367087.1 | - | 3 - 4 | 170 | 62 | |
CV815254.1 | - | 3 - 4 | 147 | 62 | |
DB452048.1 | - | 3 - 4 | 292 | 62 |