The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004329.2 | NP_004320.2 | 5 - 6 | 881 | 78 |
XM_011540103.2 | XP_011538405.1 | 5 - 6 | 778 | 78 | |
XM_011540104.2 | XP_011538406.1 | 6 - 7 | 945 | 78 | |
GenBank mRNA | AB451405.1 | - | 3 - 4 | 333 | 78 |
AK291764.1 | - | 5 - 6 | 750 | 78 | |
BC028383.1 | - | 5 - 6 | 875 | 78 | |
Z22535.1 | - | 5 - 6 | 642 | 78 |