The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006092.2 | NP_006083.1 | 2 - 3 | 305 | 159 |
XM_005249568.1 | XP_005249625.1 | 3 - 4 | 360 | 159 | |
XM_006715633.2 | XP_006715696.1 | 3 - 4 | 397 | 159 | |
XM_011515079.1 | XP_011513381.1 | 4 - 5 | 543 | 159 | |
XM_011515080.2 | XP_011513382.1 | 2 - 3 | 219 | 159 | |
XM_011515083.1 | XP_011513385.1 | 3 - 4 | 360 | 159 | |
XM_011515084.1 | XP_011513386.1 | 3 - 4 | 360 | 159 | |
XM_011515085.1 | XP_011513387.1 | 3 - 4 | 360 | 159 | |
XM_011515087.1 | XP_011513389.1 | 3 - 4 | 360 | 159 | |
XM_011515088.2 | XP_011513390.1 | 3 - 4 | 360 | 159 | |
XM_017011674.1 | XP_016867163.1 | 3 - 4 | 360 | 159 | |
XM_017011675.1 | XP_016867164.1 | 2 - 3 | 266 | 159 | |
GenBank mRNA | AF113925.1 | - | 2 - 3 | 206 | 159 |
AF126484.1 | - | 2 - 3 | 233 | 159 | |
AK023969.1 | - | 2 - 3 | 266 | 159 | |
AK300367.1 | - | 2 - 3 | 224 | 159 | |
AY128682.1 | - | 2 - 3 | 164 | 159 | |
AY128684.1 | - | 3 - 4 | 240 | 159 | |
AY128685.1 | - | 3 - 4 | 305 | 159 | |
AY128686.1 | - | 3 - 4 | 254 | 159 | |
BC040339.1 | - | 2 - 3 | 220 | 159 | |
CV569884.1 | - | 2 - 3 | 305 | 159 |