The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005249568.1 | XP_005249625.1 | 2 - 3 | 219 | 84 |
XM_011515079.1 | XP_011513381.1 | 3 - 4 | 402 | 84 | |
XM_011515081.2 | XP_011513383.1 | 2 - 3 | 317 | 84 | |
XM_011515083.1 | XP_011513385.1 | 2 - 3 | 219 | 84 | |
XM_011515084.1 | XP_011513386.1 | 2 - 3 | 219 | 84 | |
XM_011515085.1 | XP_011513387.1 | 2 - 3 | 219 | 84 | |
XM_011515087.1 | XP_011513389.1 | 2 - 3 | 219 | 84 | |
XM_011515088.2 | XP_011513390.1 | 2 - 3 | 219 | 84 | |
XM_017011674.1 | XP_016867163.1 | 2 - 3 | 219 | 84 | |
GenBank mRNA | AY128686.1 | - | 2 - 3 | 113 | 84 |