Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZFPL1
Entrez Gene ID
7542
Gene Name
zinc finger protein like 1
Gene Aliases
D11S750, MCG4
Chromosome Location
Chr.11: 65084222 - 65088402 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006782.3 NP_006773.2 1 - 2 158 57
GenBank mRNA AF001891.1 - 1 - 2 140 57
AF030291.1 - 1 - 2 145 57
AK291964.1 - 1 - 2 139 57
AK308740.1 - 1 - 2 129 57
AK316044.1 - 1 - 2 138 57
AU279924.1 - 1 - 2 158 57
BC012530.2 - 1 - 2 93 57
BC012814.2 - 1 - 2 93 57

View Full Product Details