Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PARK2
Entrez Gene ID
5071
Gene Name
parkin RBR E3 ubiquitin protein ligase
Gene Aliases
AR-JP, LPRS2, PDJ, PRKN
Chromosome Location
Chr.6: 161347417 - 162727802 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004562.2 NP_004553.2 2 - 3 306 65
NM_013987.2 NP_054642.2 2 - 3 306 65
XM_011535863.1 XP_011534165.1 2 - 3 306 65
XM_017010908.1 XP_016866397.1 2 - 3 1220 65
GenBank mRNA AB009973.1 - 2 - 3 273 65
AB245403.1 - 2 - 3 264 65
AF381284.1 - 1 - 2 172 65
AK292590.1 - 2 - 3 267 65
BC022014.2 - 2 - 3 270 65
DB023187.1 - 2 - 3 306 65
EF375726.1 - 1 - 2 172 65
GU345838.1 - 1 - 2 189 65
GU345839.1 - 2 - 3 189 65
GU345840.1 - 1 - 2 189 65
GU361466.1 - 1 - 2 189 65
GU361467.1 - 1 - 2 189 65
GU361469.1 - 2 - 3 286 65
GU361470.1 - 2 - 3 287 65
KC357595.1 - 4 - 5 384 65
KJ534911.1 - 1 - 2 172 65
KJ534912.1 - 1 - 2 172 65
KU178240.1 - 1 - 2 172 65
KU178241.1 - 1 - 2 172 65
KU178242.1 - 1 - 2 172 65

View Full Product Details