The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004562.2 | NP_004553.2 | 7 - 8 | 1007 | 83 |
NM_013987.2 | NP_054642.2 | 6 - 7 | 923 | 83 | |
NM_013988.2 | NP_054643.2 | 4 - 5 | 560 | 83 | |
XM_011535863.1 | XP_011534165.1 | 7 - 8 | 1004 | 83 | |
XM_017010908.1 | XP_016866397.1 | 7 - 8 | 1921 | 83 | |
XM_017010909.1 | XP_016866398.1 | 6 - 7 | 801 | 83 | |
GenBank mRNA | AB009973.1 | - | 7 - 8 | 974 | 83 |
AB245403.1 | - | 8 - 9 | 965 | 83 | |
AF381282.1 | - | 5 - 6 | 632 | 83 | |
AF381284.1 | - | 5 - 6 | 751 | 83 | |
AK292590.1 | - | 7 - 8 | 968 | 83 | |
AK294684.1 | - | 2 - 3 | 213 | 83 | |
EF375726.1 | - | 6 - 7 | 873 | 83 | |
GU345837.1 | - | 6 - 7 | 726 | 83 | |
GU345838.1 | - | 5 - 6 | 768 | 83 | |
GU345839.1 | - | 6 - 7 | 806 | 83 | |
GU345840.1 | - | 6 - 7 | 890 | 83 | |
GU357501.1 | - | 4 - 5 | 363 | 83 | |
GU361467.1 | - | 5 - 6 | 806 | 83 | |
GU361469.1 | - | 7 - 8 | 987 | 83 | |
GU361470.1 | - | 7 - 8 | 988 | 83 | |
KC357595.1 | - | 9 - 10 | 1085 | 83 | |
KC774171.1 | - | 5 - 6 | 740 | 83 | |
KJ534911.1 | - | 6 - 7 | 873 | 83 | |
KJ534912.1 | - | 6 - 7 | 873 | 83 | |
KU178241.1 | - | 6 - 7 | 873 | 83 | |
KU178242.1 | - | 6 - 7 | 870 | 83 |