The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004562.2 | NP_004553.2 | 9 - 10 | 1221 | 96 |
NM_013987.2 | NP_054642.2 | 8 - 9 | 1137 | 96 | |
NM_013988.2 | NP_054643.2 | 6 - 7 | 774 | 96 | |
XM_011535863.1 | XP_011534165.1 | 9 - 10 | 1218 | 96 | |
XM_017010908.1 | XP_016866397.1 | 9 - 10 | 2135 | 96 | |
XM_017010909.1 | XP_016866398.1 | 8 - 9 | 1015 | 96 | |
GenBank mRNA | AB009973.1 | - | 9 - 10 | 1188 | 96 |
AB245403.1 | - | 10 - 11 | 1179 | 96 | |
AF381282.1 | - | 7 - 8 | 846 | 96 | |
AF381284.1 | - | 7 - 8 | 965 | 96 | |
AF381286.1 | - | 2 - 3 | 223 | 96 | |
AK292590.1 | - | 9 - 10 | 1182 | 96 | |
AK294684.1 | - | 4 - 5 | 427 | 96 | |
GU345837.1 | - | 8 - 9 | 940 | 96 | |
GU345838.1 | - | 7 - 8 | 982 | 96 | |
GU345839.1 | - | 8 - 9 | 1020 | 96 | |
GU357501.1 | - | 6 - 7 | 577 | 96 | |
GU357502.1 | - | 4 - 5 | 515 | 96 | |
GU361466.1 | - | 6 - 7 | 920 | 96 | |
GU361469.1 | - | 9 - 10 | 1201 | 96 | |
GU361470.1 | - | 9 - 10 | 1202 | 96 | |
GU361471.1 | - | 3 - 4 | 276 | 96 | |
KC357595.1 | - | 11 - 12 | 1299 | 96 | |
KC774171.1 | - | 7 - 8 | 954 | 96 | |
KJ534911.1 | - | 8 - 9 | 1087 | 96 | |
KJ534912.1 | - | 8 - 9 | 1087 | 96 |