The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001322923.1 | NP_001309852.1 | 4 - 5 | 616 | 73 |
NM_001322924.1 | NP_001309853.1 | 3 - 4 | 544 | 73 | |
NM_052862.3 | NP_443094.3 | 5 - 6 | 662 | 73 | |
GenBank mRNA | AF545852.1 | - | 5 - 6 | 806 | 73 |
AL137762.1 | - | 5 - 6 | 583 | 73 | |
AL833211.1 | - | 3 - 4 | 352 | 73 | |
AY530954.1 | - | 4 - 5 | 475 | 73 | |
BC101536.1 | - | 5 - 6 | 499 | 73 | |
BC101562.1 | - | 5 - 6 | 499 | 73 | |
BC143428.1 | - | 4 - 5 | 409 | 73 |