Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LIMD1
Entrez Gene ID
8994
Gene Name
LIM domains containing 1
Gene Aliases
-
Chromosome Location
Chr.3: 45594181 - 45681263 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014240.2 NP_055055.1 4 - 5 1696 95
GenBank mRNA AJ132408.1 - 4 - 5 1668 95
BC117236.1 - 4 - 5 1696 95
BC117238.1 - 4 - 5 1696 95

View Full Product Details